ADVH49B834DC AChemBlock

(R)-3,3-difluoro-4-hydroxy-N-((S)-1-phenylethyl)piperidine-1-carboxamide

  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-3-difluoro-4-hydroxy-n-s-1-phenylethyl-piperidine-1-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88854
Availability details for
Quantity
  • Availability
  • Add to Cart