ADVH49B834DD AChemBlock

(R)-1-((R)-3,3-difluoro-4-hydroxypiperidin-1-yl)-2-hydroxy-2-phenylethan-1-one

  • Molecular Weight : 271.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-r-3-3-difluoro-4-hydroxypiperidin-1-yl-2-hydroxy-2-phenylethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88855
Availability details for
Quantity
  • Availability
  • Add to Cart