ADVH49B834E0 AChemBlock

(S)-3-amino-4-(3,5-difluorophenyl)butanoic acid hydrochloride

  • Molecular Weight : 251.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-amino-4-3-5-difluorophenyl-butanoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89105
Availability details for
Quantity
  • Availability
  • Add to Cart