ADVH49B834E1 AChemBlock

(R)-3-amino-4-(3,5-difluorophenyl)butanoic acid hydrochloride

  • Molecular Weight : 251.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-amino-4-3-5-difluorophenyl-butanoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89106
Availability details for
Quantity
  • Availability
  • Add to Cart