ADVH49B834E3 AChemBlock

(R)-3-((tert-butoxycarbonyl)amino)-4-(3,5-difluorophenyl)butanoic acid

  • Molecular Weight : 315.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-tert-butoxycarbonyl-amino-4-3-5-difluorophenyl-butanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89108
Availability details for
Quantity
  • Availability
  • Add to Cart