ADVH49B834E6 AChemBlock

tert-butyl(1H-pyrazol-4-yl)carbamate

  • Cas Number : 130106-42-2
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1h-pyrazol-4-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89135
Availability details for
Quantity
  • Availability
  • Add to Cart