ADVH49B834E8 AChemBlock

N-acetyl-S-(4-methylbenzoyl)-D-cysteine

  • Cas Number : 1695562-36-7
  • Molecular Weight : 281.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-acetyl-s-4-methylbenzoyl-d-cysteine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89189
Availability details for
Quantity
  • Availability
  • Add to Cart