ADVH49B834E9 AChemBlock

2-methoxy-N-(2-methoxyphenyl)benzimidamide

  • Cas Number : 524923-93-1
  • Molecular Weight : 256.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-n-2-methoxyphenyl-benzimidamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89214
Availability details for
Quantity
  • Availability
  • Add to Cart