ADVH49B834EA AChemBlock

butane-1,4-diyl bis(3,4-dimethoxybenzoate)

  • Cas Number : 72782-65-1
  • Molecular Weight : 418.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/butane-1-4-diyl-bis-3-4-dimethoxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89226
Availability details for
Quantity
  • Availability
  • Add to Cart