ADVH49B834EF AChemBlock

4-bromo-1-(tert-butyl)-2-ethyl-2,3,6,7-tetrahydro-1H-1l4-pyrrolo[2,3-b]pyridine-1,2-dicarboxylate

  • Cas Number : 1269449-33-3
  • Molecular Weight : 372.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1-tert-butyl-2-ethyl-2-3-6-7-tetrahydro-1h-1l4-pyrrolo-2-3-b-pyridine-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89553
Availability details for
Quantity
  • Availability
  • Add to Cart