ADVH49B834F1 AChemBlock

Fmoc-D-phe(CBZ-4-Amino)-OH

  • Molecular Weight : 536.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmoc-d-phe-cbz-4-amino-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89628
Availability details for
Quantity
  • Availability
  • Add to Cart