ADVH49B834F2 AChemBlock

CBZ-D-

  • Molecular Weight : 351.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/cbz-d-hoglu-otbu-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89640
Availability details for
Quantity
  • Availability
  • Add to Cart