ADVH49B834F5 AChemBlock

Cycloguanil dihydrochloride

  • Cas Number : 25332-47-2
  • Molecular Weight : 324.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cycloguanil-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89691
Availability details for
Quantity
  • Availability
  • Add to Cart