ADVH49B834F6 AChemBlock

5-(tert-butyl)-3-(2,4-dichloro-5-hydroxyphenyl)-1,3,4-oxadiazol-2(3H)-one

  • Molecular Weight : 303.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-tert-butyl-3-2-4-dichloro-5-hydroxyphenyl-1-3-4-oxadiazol-2-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88670
Availability details for
Quantity
  • Availability
  • Add to Cart