ADVH49B834F8 AChemBlock

(1-methyl-1H-indol-2-yl)methanol

  • Cas Number : 1485-22-9
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-1h-indol-2-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88557
Availability details for
Quantity
  • Availability
  • Add to Cart