ADVH49B834FA AChemBlock

3-amino-5-chloro-N-cyclopropyl-4-methyl-6-(methylthio)thieno[2,3-b]pyridine-2-carboxamide

  • Molecular Weight : 327.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-5-chloro-n-cyclopropyl-4-methyl-6-methylthio-thieno-2-3-b-pyridine-2-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88632
Availability details for
Quantity
  • Availability
  • Add to Cart