ADVH49B834FB AChemBlock

1-benzyl 2-methyl 1H-indole-1,2-dicarboxylate

  • Molecular Weight : 309.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-2-methyl-1h-indole-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88675
Availability details for
Quantity
  • Availability
  • Add to Cart