ADVH49B834FE AChemBlock

(S)-3,3'-diiodo-2,2'-dimethoxy-1,1'-binaphthalene

  • Molecular Weight : 566.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-3-diiodo-2-2-dimethoxy-1-1-binaphthalene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88678
Availability details for
Quantity
  • Availability
  • Add to Cart