ADVH49B83500 AChemBlock

6-iodo-3-(pyridin-2-yl)-3,4-dihydro-5-oxa-1,2a-diazaacenaphthylen-2(1H)-one

  • Molecular Weight : 379.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-iodo-3-pyridin-2-yl-3-4-dihydro-5-oxa-1-2a-diazaacenaphthylen-2-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88633
Availability details for
Quantity
  • Availability
  • Add to Cart