ADVH49B83501 AChemBlock

2,6-bis(4-(tert-butyl)-2,6-diisopropylphenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

  • Molecular Weight : 781.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-6-bis-4-tert-butyl-2-6-diisopropylphenyl-4-hydroxydinaphtho-2-1-d-1-2-f-1-3-2-dioxaphosphepine-4-oxide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88685
Availability details for
Quantity
  • Availability
  • Add to Cart