ADVH49B83506 AChemBlock

(3-(3-amino-2,6-difluorobenzoyl)-5-(2-cyclopropylpyrimidin-5-yl)-1H-pyrrolo[2,3-b]pyridin-1-yl)(2,6-dichlorophenyl)methanone

  • Molecular Weight : 564.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-3-amino-2-6-difluorobenzoyl-5-2-cyclopropylpyrimidin-5-yl-1h-pyrrolo-2-3-b-pyridin-1-yl-2-6-dichlorophenyl-methanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88636
Availability details for
Quantity
  • Availability
  • Add to Cart