ADVH49B83507 AChemBlock

4,4'-((1E,1'E)-(1-phenyl-1H-pyrazole-3,5-diyl)bis(ethene-2,1-diyl))bis(2-methoxyphenol)

  • Cas Number : 1019110-87-2
  • Molecular Weight : 440.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-1e-1-e-1-phenyl-1h-pyrazole-3-5-diyl-bis-ethene-2-1-diyl-bis-2-methoxyphenol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88691
Availability details for
Quantity
  • Availability
  • Add to Cart