ADVH49B8350A AChemBlock

tert-butyl (R)-4-(2-oxo-2-(2-oxo-4-phenyloxazolidin-3-yl)ethyl)piperidine-1-carboxylate

  • Cas Number : 1159845-42-7
  • Molecular Weight : 388.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-r-4-2-oxo-2-2-oxo-4-phenyloxazolidin-3-yl-ethyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88697
Availability details for
Quantity
  • Availability
  • Add to Cart