ADVH49B8350C AChemBlock

ethyl 1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate

  • Cas Number : 1159982-41-8
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-benzyl-1-2-5-6-tetrahydropyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88699
Availability details for
Quantity
  • Availability
  • Add to Cart