ADVH49B8350D AChemBlock

tert-butyl 3-(2-(tert-butoxy)-2-oxoethyl)piperidine-1-carboxylate

  • Cas Number : 1159982-66-7
  • Molecular Weight : 299.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-2-tert-butoxy-2-oxoethyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88701
Availability details for
Quantity
  • Availability
  • Add to Cart