ADVH49B8350E AChemBlock

5-hydroxy-5-(trifluoromethyl)dihydrofuran-2(3H)-one

  • Cas Number : 117192-54-8
  • Molecular Weight : 170.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-hydroxy-5-trifluoromethyl-dihydrofuran-2-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88702
Availability details for
Quantity
  • Availability
  • Add to Cart