ADVH49B83513 AChemBlock

2-aminoisoindoline-1,3-dione hydrochloride

  • Cas Number : 1198286-64-4
  • Molecular Weight : 198.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-aminoisoindoline-1-3-dione-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88707
Availability details for
Quantity
  • Availability
  • Add to Cart