ADVH49B83515 AChemBlock

tert-butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate

  • Cas Number : 1215071-17-2
  • Molecular Weight : 235.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-3-difluoro-4-oxopiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88712
Availability details for
Quantity
  • Availability
  • Add to Cart