ADVH49B83518 AChemBlock

tert-butyl 2-(((benzyloxy)carbonyl)amino)-7-azabicyclo[2.2.1]heptane-7-carboxylate

  • Cas Number : 1221818-02-5
  • Molecular Weight : 346.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-benzyloxy-carbonyl-amino-7-azabicyclo-2-2-1-heptane-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88715
Availability details for
Quantity
  • Availability
  • Add to Cart