ADVH49B83519 AChemBlock

4-bromo-1-propyl-1,2,3,6-tetrahydropyridine

  • Cas Number : 1221818-50-3
  • Molecular Weight : 204.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1-propyl-1-2-3-6-tetrahydropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88716
Availability details for
Quantity
  • Availability
  • Add to Cart