ADVH49B8351D AChemBlock

tert-butyl 4-(cyclopropylmethyl)piperidine-1-carboxylate

  • Cas Number : 1241725-67-6
  • Molecular Weight : 239.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-cyclopropylmethyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88720
Availability details for
Quantity
  • Availability
  • Add to Cart