ADVH49B8351E AChemBlock

2-(1-(tert-butoxycarbonyl)-4-hydroxypiperidin-4-yl)-2-methylpropanoic acid

  • Cas Number : 1251032-53-7
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-1-tert-butoxycarbonyl-4-hydroxypiperidin-4-yl-2-methylpropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88722
Availability details for
Quantity
  • Availability
  • Add to Cart