ADVH49B83522 AChemBlock

tert-butyl 3-amino-4-fluoropyrrolidine-1-carboxylate

  • Cas Number : 1260848-87-0
  • Molecular Weight : 204.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-amino-4-fluoropyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88726
Availability details for
Quantity
  • Availability
  • Add to Cart