ADVH49B83525 AChemBlock

(E)-1,1'-dibenzyl-5,5',6,6'-tetrahydro-1'H,2H-[3,4'-bipyridinylidene]-2,2',4(1H,3'H)-trione

  • Cas Number : 1283734-84-8
  • Molecular Weight : 388.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/e-1-1-dibenzyl-5-5-6-6-tetrahydro-1-h-2h-3-4-bipyridinylidene-2-2-4-1h-3-h-trione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88729
Availability details for
Quantity
  • Availability
  • Add to Cart