ADVH49B83526 AChemBlock

6-fluoro-1-methyl-3-(piperidin-4-yl)-1H-indazole hydrochloride

  • Cas Number : 129014-50-2
  • Molecular Weight : 269.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-fluoro-1-methyl-3-piperidin-4-yl-1h-indazole-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88730
Availability details for
Quantity
  • Availability
  • Add to Cart