ADVH49B83527 AChemBlock

1-(tert-butyl) 4-methyl 3-fluoropiperidine-1,4-dicarboxylate

  • Cas Number : 1303974-48-2
  • Molecular Weight : 261.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-4-methyl-3-fluoropiperidine-1-4-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88731
Availability details for
Quantity
  • Availability
  • Add to Cart