ADVH49B83530 AChemBlock

tert-butyl 3-methylpiperidine-1-carboxylate

  • Cas Number : 146337-18-0
  • Molecular Weight : 199.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-methylpiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88740
Availability details for
Quantity
  • Availability
  • Add to Cart