ADVH49B83531 AChemBlock

(R)-4-((3R,5R,8S,9S,10R,13R,14S,17R,E)-6-ethylidene-3-hydroxy-10,13-dimethyl-7-oxohexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid

  • Cas Number : 1516887-33-4
  • Molecular Weight : 416.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-4-3r-5r-8s-9s-10r-13r-14s-17r-e-6-ethylidene-3-hydroxy-10-13-dimethyl-7-oxohexadecahydro-1h-cyclopenta-a-phenanthren-17-yl-pentanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88743
Availability details for
Quantity
  • Availability
  • Add to Cart