ADVH49B83536 AChemBlock

tert-butyl 4-((4-fluorophenyl)(hydroxy)methyl)piperidine-1-carboxylate

  • Cas Number : 160296-41-3
  • Molecular Weight : 309.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-4-fluorophenyl-hydroxy-methyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88748
Availability details for
Quantity
  • Availability
  • Add to Cart