ADVH49B83537 AChemBlock

tert-butyl 4-(1H-indol-1-yl)piperidine-1-carboxylate

  • Cas Number : 170364-89-3
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-1h-indol-1-yl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88750
Availability details for
Quantity
  • Availability
  • Add to Cart