ADVH49B8353B AChemBlock

benzyl 4-((tert-butoxycarbonyl)amino)-3-fluoropiperidine-1-carboxylate

  • Cas Number : 1820676-07-0
  • Molecular Weight : 352.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzyl-4-tert-butoxycarbonyl-amino-3-fluoropiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88754
Availability details for
Quantity
  • Availability
  • Add to Cart