ADVH49B8353D AChemBlock

tert-butyl 4-fluoro-3-methylenepiperidine-1-carboxylate

  • Cas Number : 1823905-84-5
  • Molecular Weight : 215.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-fluoro-3-methylenepiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88756
Availability details for
Quantity
  • Availability
  • Add to Cart