ADVH49B8353E AChemBlock

2,2-difluoro-3,3-dihydroxycyclohexan-1-one

  • Cas Number : 183742-86-1
  • Molecular Weight : 166.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluoro-3-3-dihydroxycyclohexan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88757
Availability details for
Quantity
  • Availability
  • Add to Cart