ADVH49B83542 AChemBlock

tert-butyl (3R,4S)-4-fluoro-3-(hydroxymethyl)piperidine-1-carboxylate

  • Cas Number : 1903839-17-7
  • Molecular Weight : 233.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-4s-4-fluoro-3-hydroxymethyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88762
Availability details for
Quantity
  • Availability
  • Add to Cart