ADVH49B83545 AChemBlock

(R)-2-((1s,4S)-4-(6-fluoroquinolin-4-yl)cyclohexyl)propanoic acid

  • Cas Number : 1923836-34-3
  • Molecular Weight : 301.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-1s-4s-4-6-fluoroquinolin-4-yl-cyclohexyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88765
Availability details for
Quantity
  • Availability
  • Add to Cart