ADVH49B83546 AChemBlock

(R)-3-(2-(4-(6-fluoroquinolin-4-yl)cyclohexyl)acetyl)-4-phenyloxazolidin-2-one

  • Cas Number : 1923836-68-3
  • Molecular Weight : 432.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-2-4-6-fluoroquinolin-4-yl-cyclohexyl-acetyl-4-phenyloxazolidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88766
Availability details for
Quantity
  • Availability
  • Add to Cart