ADVH49B83548 AChemBlock

(E)-3-((2-(2,4-dimethylphenyl)-2-(2,2,2-trifluoroacetyl)hydrazineylidene)methyl)benzoic acid

  • Cas Number : 1980780-69-5
  • Molecular Weight : 364.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/e-3-2-2-4-dimethylphenyl-2-2-2-2-trifluoroacetyl-hydrazineylidene-methyl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88768
Availability details for
Quantity
  • Availability
  • Add to Cart