ADVH49B83549 AChemBlock

ethyl 1-benzyl-2,4-dioxopiperidine-3-carboxylate

  • Cas Number : 198417-15-1
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-benzyl-2-4-dioxopiperidine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88769
Availability details for
Quantity
  • Availability
  • Add to Cart