ADVH49B8354C AChemBlock

tert-butyl 4-(2-ethoxy-2-oxoethyl)piperazine-1-carboxylate

  • Cas Number : 209667-59-4
  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-2-ethoxy-2-oxoethyl-piperazine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88773
Availability details for
Quantity
  • Availability
  • Add to Cart