ADVH49B8354D AChemBlock

methyl 1-benzylpyrrolidine-3-carboxylate oxalate

  • Cas Number : 2101206-24-8
  • Molecular Weight : 309.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-benzylpyrrolidine-3-carboxylate-oxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88774
Availability details for
Quantity
  • Availability
  • Add to Cart